Learn More
Medchemexpress LLC 7-Methylcoumarin | 2445-83-2 | 10 G
SDP
Supplier:  Medchemexpress LLC HYN509210G
Encompass_Preferred
7-Methylcoumarin is a coumarin derivative with potent hepatoprotective and antioxidant properties, acting as a mechanism-based inhibitor for CYP2A6. It significantly reduces alanine aminotransferase (ALT), aspartate aminotransferase (AST), and serum bilirubin (TB) in rats with CCl4-induced liver damage. It also restores total protein (TP) and albumin (TA) levels in serum, prevents oxidative stress, and can decrease the mitotic activity of *A. sativum* promeristem.
  • Acts as a mechanism-based inhibitor for human CYP2A6.
  • Can induce chromosomal aberrations in *A. sativum* promeristem.
  • Restores elevated alanine aminotransferase (ALT) and aspartate aminotransferase (AST) levels in rats.
  • Restores antioxidant enzyme and TBARS levels.
  • Restores serum bilirubin (TB) and total protein (TP) levels.
  • Reduces hepatic lesions induced by CCl4.
Catalog No. 50-004-19597