Learn More
Cayman Chemical 7-MethylgunosIn-d3 1mg
SDP
Supplier:  Cayman Chemical 363361 MG
Encompass_Preferred
An internal standard for the quantification of 7-methylguanosine by GC- or LC-MS
Catalog No. 50-291-2175