Learn More
Cayman Chemical 7-MethylgunosIn-d3 500ug
SDP
Supplier:  Cayman Chemical 36336500 UG
Encompass_Preferred
An internal standard for the quantification of 7-methylguanosine by GC- or LC-MS
Catalog No. 50-291-2166