Learn More
Cayman Chemical 7-NItroIndazole 250mg
SDP
Supplier:  Cayman Chemical 81340250 MG
Encompass_Preferred
A non-selective inhibitor of NOS isoforms in vitro (IC50s = 0.71, 0.78, and 5.8 µM for rat nNOS, bovine eNOS, and rat iNOS, respectively); shows good anti-nociceptive effects without affecting blood pressure via inhibition of eNOS in vivo
Catalog No. 50-289-4044