Learn More
Medchemexpress LLC 7-O-(amino-PEG4)-paclitaxel | 00-00-0 | 99.8% | 1101.19 g·mol⁻¹ | C58H72N2O19 | 5 MG

Supplier: Medchemexpress LLC HY1411475MG
7-O-(Amino-PEG4)-paclitaxel is a PEG-based drug-linker conjugate that incorporates a paclitaxel moiety with a terminal amino group. It is intended for research use in the synthesis of antibody-drug conjugates (ADCs) and as a PROTAC linker, providing a convenient reactive handle for downstream coupling.
- Contains a paclitaxel moiety with a terminal primary amine.
- Amino group reactive with carboxylic acids, activated NHS esters, and carbonyls.
- Suitable for ADC synthesis and PROTAC construction.
- High molecular weight suitable for linker applications (approx. 1101.19 g·mol⁻¹).
- Provided in small research pack sizes with reported purity up to 99.8%.
- Store powder at -20°C; store in solvent at -80°C for long-term stability.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.