Learn More
Medchemexpress LLC 7-Octynoic acid | 10297-09-3 | 98.0% | C8H12O2 | 500 MG
SDP
Supplier:  Medchemexpress LLC HY69220500MG
Encompass_Preferred
7-Octynoic acid is a PROTAC linker (compound 42) that facilitates the synthesis of PROTACs. PROTACs are designed to harness the intracellular ubiquitin-proteasome system, utilizing two distinct ligands-one targeting an E3 ubiquitin ligase and the other the target protein-to achieve selective degradation of target proteins. Additionally, this compound serves as a click chemistry reagent, featuring an Alkyne group capable of participating in copper-catalyzed azide-alkyne cycloaddition (CuAAc) reactions with molecules containing Azide groups.
  • Facilitates PROTAC synthesis
  • Connects ligands for E3 ubiquitin ligase and target proteins
  • Leverages the ubiquitin-proteasome system for selective protein degradation
  • Acts as a click chemistry reagent
  • Contains an alkyne group for CuAAc reactions
Catalog No. 50-004-33673