Learn More
Enzo Life Sciences 7-Oxostaurosporine (1mg). CAS: 125035-83-8
SDP
Supplier:  Enzo Life Sciences ALX380210M001
Encompass_Preferred
Antibiotic. Potent inhibitor of protein kinase C (PKC). Inhibits the cell cycle at the G2 stage. Alternative name: Antibiotic BMY 41950, Antibiotic RK 1409, LCM76-L. Purity: ≥98% (HPLC). Solubility: Soluble in DMSO, dimethyl formamide, methanol or 100% ethanol; slightly soluble in water. Long Term Storage: -20°C.
Catalog No. 50-200-8974