Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 7-pentadecanol | 4104-59-0 | MFCD00046732 | 98.7% | 228.41 g/mol | C15H32O | 1 G
SDP

Supplier:  Medchemexpress LLC HYW5948331G

Encompass_Preferred

7-Pentadecanol is a long-chain fatty alcohol with a terminal hydroxyl group offered as a research reagent. It is characterized by analytical data supporting identity and purity and is commonly used in biochemical and lipid research.

  • Purity: 98.7% (GC).
  • Molecular formula: C15H32O; molecular weight: 228.41 g/mol.
  • Appearance: white to off-white solid.
  • Analytical data: 1H NMR consistent with structure; optical rotation 0.467° (C=0.01 g/ml, CHCl3).
  • Storage: room temperature (3 years); in solvent at -80°C (2 years) or -20°C (1 year).
  • Intended use: for biochemical research applications.

Catalog No. 50-002-84006


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.