Learn More
Medchemexpress LLC 7-Prenyloxycoumarin | 10387-50-5 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYN70235MG
Encompass_Preferred
7-Prenyloxycoumarin, also known as 7-O-Prenylumbelliferone, is a secondary metabolite. It is derived from the endophytic fungus of *Annulohypoxylon ilanense*. This compound is supplied as a white to off-white solid with a molecular weight of 230.26 and a formula of C14H14O3. It demonstrates varying levels of cytotoxicity against several human cancer cell lines, including BT-549 and LoVo, with IC50 values of 37 μM and 30 μM respectively, while showing higher IC50 values (>100 μM) against A549, PC-3, SK-MEL-28, and U-373MG ATCC cell lines. 7-Prenyloxycoumarin is intended for research use only.
  • Derived from natural sources
  • Exhibits antiproliferative activity against certain cancer cell lines
  • Provided as a high-purity solid (99.87%)
  • Available in various quantities for research needs
  • Detailed solubility and storage information provided
Catalog No. 50-004-19937