Learn More
Electron Microscopy Sciences ProScope 7' Pro Slim Kit - Hawkeye Precision Borescope
SDP
Supplier:  Electron Microscopy Sciences 6835303
Encompass
Proscope Hawkeye Pro Flexible borescopes incorporate an 18,000 fiber, acid-leached imaging bundle, delivering truly excellent image resolution, sharpness, detail, and contrast. For use with the ProScope Micro Mobile digital microscope.
Catalog No. 50-190-2715
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.