Learn More
Cayman Chemical 7 S 10 S-DIHOME 5mg
SDP
Supplier:  Cayman Chemical 334475 MG
Encompass_Preferred
An antibacterial hydroxy fatty acid; active against various food-borne pathogenic bacteria (MIC50s = 31.3-250 UG/ml), as well as various plant pathogenic bacteria (MIC90s = 125-500 UG/ml, respectively)
Catalog No. 50-294-8713