Learn More
Cayman Chemical 7 Z 10 Z 13 ZHexadecatrienoic
SDP
Supplier:  Cayman Chemical 100051565 MG
Encompass_Preferred
An ω-3 PUFA; a precursor in the biosynthesis of jasmonic acid in A. thaliana leaves
Catalog No. 50-288-9035