Learn More
Medchemexpress LLC 7(z),11(z)-heptacosadiene | 100462-58-6 | MFCD11976900 | 99.9% | 376.70 g/mol | C27H52 | 5 MG
SDP
Supplier:  Medchemexpress LLC HY1125275MG
Encompass_Preferred
7(Z),11(Z)-Heptacosadiene is a long-chain, unsaturated hydrocarbon diene provided for research use. It is employed in biochemical and metabolic studies, including semiochemical and pheromone research, and is supplied in small, quantified packs with a high reported purity.
  • Long-chain unsaturated diene (C27H52).
  • Molecular weight 376.70 g/mol.
  • High purity, reported as 99.9%.
  • Provided in small research quantities (for example, 5 mg).
  • Intended for biochemical and metabolic research applications.
  • Commonly used in insect pheromone and semiochemical studies.
Catalog No. 50-002-62432