Learn More
Cayman Chemical 7 Z 11 ZHeptacosadiene
SDP
Supplier:  Cayman Chemical 100125675 MG
Encompass_Preferred
Female mating pheromone of D. melanogaster; elicits wing vibrations in male D. melanogaster flies at >100 ng
Catalog No. 50-288-8589