Learn More
ABclonal Technology 700 Rabbit anti- CD162/PSGL-1
SDP
Supplier:  ABclonal Technology A25947500T
Encompass_Preferred
Involved in leukocyte adhesive activation Acts upstream of or within leukocyte tethering or rolling Predicted to be located in plasma membrane raft and uropod Predicted to be active in plasma membrane Is expressed in brain and thymus primordium Human ortholog(s) of this gene implicated in carotid artery disease Orthologous to human SELPLG (selectin P ligand)
Catalog No. 50-000-12597