Learn More
ABclonal Technology 700 Rabbit anti-Human CD1c
SDP
Supplier:  ABclonal Technology A26842500T
Encompass_Preferred
This gene encodes a member of the CD1 family of transmembrane glycoproteins which are structurally related to the major histocompatibility complex (MHC) proteins and form heterodimers with beta-2-microglobulin The CD1 proteins mediate the presentation of primarily lipid and glycolipid antigens of self or microbial origin to T cells The human genome contains five CD1 family genes organized in a cluster on chromosome 1 The CD1 family members are thought to differ in their cellular localization and specificity for particular lipid ligands The protein encoded by this gene is broadly distributed throughout the endocytic system via a tyrosine-based motif in the cytoplasmic tail Alternatively spliced transcript variants of this gene have been observed but their full-length nature is not known
Catalog No. 50-000-08421