Learn More
Sino Biological Recombinant Canine Ephrin B2 Protein (His Tag)
Supplier:  Sino Biological 70073D08H100
Encompass_Preferred
A DNA sequence encoding the canine EFNB2 (NP_001107768.1) (Met1-Ala229) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-3403