Learn More
Sino Biological Recombinant Canine IL-11RA/IL-11R alpha Protein (hFc Tag)
Supplier:  Sino Biological 70078D02H200
Encompass_Preferred
A DNA sequence encoding the canine IL11RA (XP_538710.4) (Met1-Leu363) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3407