Learn More
Sino Biological Recombinant Canine IL-11RA/IL-11R alpha Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 70078D08H100
Encompass_Preferred
A DNA sequence encoding the canine IL11RA (XP_005626799.1) (Met1-Leu363) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-3409