Learn More
Sino Biological Recombinant Canine IL-11RA/IL-11R alpha Protein
Supplier:  Sino Biological 70078DCCH100
Encompass_Preferred
A DNA sequence encoding the canine IL11RA (XP_538710.4) (Met1-Leu363) was expressed with six amino acids (LEVLFQ) at the C-terminus.
Catalog No. 50-162-3411