Learn More
Sino Biological Recombinant Canine IL-13 R alpha 2/IL13RA2 Protein (hFc Tag), HPLC-verified
Supplier:  Sino Biological 70079D02H100
Encompass_Preferred
A DNA sequence encoding the canine IL13RA2 (NP_001003075.1) (Met1-Thr338) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3413