Learn More
Sino Biological Recombinant Canine IL-13 R alpha 2/IL13RA2 Protein (His Tag), HPLC-verified
Supplier:  Sino Biological 70079D08H100
Encompass_Preferred
A DNA sequence encoding the canine IL13RA2 (Q95LF0) (Met1-Thr338) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-3415