Learn More
Sino Biological Recombinant Canine IL22RA1 Protein (hFc Tag)
Supplier:  Sino Biological 70083D02H100
Encompass_Preferred
A DNA sequence encoding the canine IL22RA1 (XP_855113.2) (Met1-Thr226) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3428