Learn More
Sino Biological Recombinant Canine IL22RA1 Protein (His Tag)
Supplier: Sino Biological 70083D08H100
A DNA sequence encoding the canine IL22RA1 (XP_855113.2) (Met1-The226) was expressed with a C-terminal polyhistidine tag.