Learn More
Sino Biological Recombinant Canine Neuregulin-1/NRG1 Protein (EGF Domain, mFc Tag)
Supplier:  Sino Biological 70084D04H100
Encompass_Preferred
A DNA sequence encoding the canine NRG1 (XP_858147.1) EGF-like domain (Ser176-Lys240) was expressed with the Fc region of mouse IgG1 at the N-terminus.
Catalog No. 50-162-3436