Learn More
Sino Biological Recombinant Canine Neuregulin-1/NRG1 Protein (ECD, His Tag)
Supplier:  Sino Biological 70084D08H100
Encompass_Preferred
A DNA sequence encoding the canine NRG1 (XP_858147.1) extracellular domain (Ser19-Lys240) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-3443