Learn More
Sino Biological Recombinant Canine Neuregulin-1/NRG1 Protein (ECD)
Supplier: Sino Biological 70084DCCH100
A DNA sequence encoding the canine NRG1 (XP_858147.1) extracellular domain (Ser19-Lys240) was expressed with five amino acids (DDDDK) at the C- terminus.