Learn More
Sino Biological Recombinant Canine TGF beta 1 Protein (Latent, His Tag)
Supplier:  Sino Biological 70087D08H20
Encompass_Preferred
A DNA sequence encoding the canine TGFB1 (XP_038300996.1) (Met1-Ser390) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3449