Learn More
Sino Biological Recombinant Canine Lymphotactin/XCL1/SCYC1 Protein (His Tag)
Supplier:  Sino Biological 70096D08B50
Encompass_Preferred
A DNA sequence encoding the canine XCL1 (XP_547477.2) (Met1-Thr112) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-3457