Learn More
Sino Biological Recombinant Canine TGF beta 2 Protein (His Tag)
Supplier:  Sino Biological 70099D08H20
Encompass_Preferred
A DNA sequence encoding the canine TGFB2 (XP_858677.1) (Met1-Ser414) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3459