Learn More
Sino Biological Recombinant Canine TrkA Protein (hFc Tag)
Supplier: Sino Biological 70101D01H100
A DNA sequence encoding the canine NTRK1 (XP_851619.3) (Pro285-Lys410) was expressed with the Fc region of human IgG1 at the N-terminus.