Learn More
Sino Biological Recombinant Canine TrkA Protein (His Tag)
Supplier:  Sino Biological 70101D07H100
Encompass_Preferred
A DNA sequence encoding the canine NTRK1 (XP_851619.3) (Pro285-Lys410) was expressed with a N-terminal polyhistidine tag.
Catalog No. 50-162-3464