Learn More
Sino Biological Recombinant Canine IL-17A Protein
Supplier: Sino Biological 70102DNAE100
A DNA sequence encoding the canine IL17A (NP_001159350.1)(Gly26-Ala155) was expressed with a N-terminal Met.