Learn More
Sino Biological Recombinant Canine IL17RD Protein (hFc Tag)
Supplier:  Sino Biological 70104D02H50
Encompass_Preferred
A DNA sequence encoding the canine IL17RD (XP_541835.3) (Ala27-Arg299) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3469