Learn More
Sino Biological Recombinant Canine IL17RD Protein (His Tag)
Supplier:  Sino Biological 70104D08H100
Encompass_Preferred
A DNA sequence encoding the canine IL17RD (XP_541835.3) (Ala27-Arg299) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-3470