Learn More
Sino Biological Recombinant Canine CD40/TNFRSF5 Protein (hFc Tag)
Supplier:  Sino Biological 70105D02H100
Encompass_Preferred
A DNA sequence encoding the canine CD40 (Q7YRL5) (Met1-Ala194) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3472