Learn More
Sino Biological Recombinant Canine CD40/TNFRSF5 Protein (His Tag)
Supplier:  Sino Biological 70105D08H100
Encompass_Preferred
A DNA sequence encoding the canine CD40 (Q7YRL5) (Met1-Ala194) was expressed with a C-terminal polyhistidine tag.
Catalog No. 50-162-3474