Learn More
Sino Biological Recombinant Canine CD40/TNFRSF5 Protein
Supplier:  Sino Biological 70105DCCH100
Encompass_Preferred
A DNA sequence encoding the canine CD40 (Q7YRL5) (Met1-Ala194) was expressed with six amino acids (LEVLFQ) at the C-terminus.
Catalog No. 50-162-3478