Learn More
Sino Biological Recombinant Canine CD3 epsilon/CD3e Protein (hFc Tag)
Supplier: Sino Biological 70106D02H100
A DNA sequence encoding the canine CD3E (NP_001003379.1) (Met1-Leu122) was expressed with the Fc region of human IgG1 at the C-terminus.