Learn More
Sino Biological Recombinant Canine CD2 Protein (hFc Tag)
Supplier:  Sino Biological 70107D02H100
Encompass_Preferred
A DNA sequence encoding the canine CD2 (XP_849219.1) (Met1-Gly204) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3482