Learn More
Sino Biological Recombinant Canine CD3 delta/CD3D Protein (hFc Tag)
Supplier:  Sino Biological 70108D02H100
Encompass_Preferred
A DNA sequence encoding the canine CD3D (XP_536556.1) (Met1-Thr103) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3486