Learn More
Sino Biological Recombinant Canine PD-L1/B7-H1 Protein (ECD, hFc Tag), HPLC-verified
Supplier:  Sino Biological 70110D02H100
Encompass_Preferred
A DNA sequence encoding the canine CD274 (NP_001278901.1) (Met1-Arg236) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3492