Learn More
Sino Biological Recombinant Canine BAFF/BLyS/TNFSF13B Protein (hFc Tag)
Supplier:  Sino Biological 70112D01H50
Encompass_Preferred
A DNA sequence encoding the canine TNFSF13B (NP_001155182.1) (Ala143-Leu292) was expressed with the Fc region of human IgG1 at the N-terminus.
Catalog No. 50-162-3495