Learn More
Sino Biological Recombinant Canine FOLR1 Protein (hFc Tag)
Supplier:  Sino Biological 70115D02H100
Encompass_Preferred
A DNA sequence encoding the canine FOLR1 (XP_851993.1) (Met1-Met228) was expressed with the Fc region of human IgG1 at the C-terminus.
Catalog No. 50-162-3500