Learn More
Sino Biological Recombinant Canine IGFBP1 Protein (His Tag)
Supplier:  Sino Biological 70254D08H100
Encompass_Preferred
A DNA sequence encoding the canine IGFBP1 (XP_005629556.1) (Met1-Ser249) was expressed with a polyhistidine tag at the C-terminus.
Catalog No. 50-162-3507