Learn More
Medchemexpress LLC 7030B-C5 PCSK9 inhibitor | 370871-42-4 | 99.78% | 359.38 | 50 MG
Supplier: Medchemexpress LLC HY16193950MG
7030B-C5 is a PCSK9 inhibitor with an IC50 of 1.61 μM. It can significantly reduce plasma cholesterol and triglyceride (TG) levels in vivo and slow the progression of atherosclerosis. This product is intended for research use only and can be used in the study of cardiovascular diseases.
- Significantly reduces plasma cholesterol and triglyceride levels
- Slows the progression of atherosclerosis
- For research use only in the study of cardiovascular diseases