Learn More
Medchemexpress LLC 7030B-C5 | 370871-42-4 | 99.8% | 359.38 | 5 MG
Supplier: Medchemexpress LLC HY1619395MG
7030B-C5 is a potent PCSK9 inhibitor (IC50=1.61 μM) that significantly reduces plasma cholesterol and triglyceride (TG) levels in vivo. It slows the progression of atherosclerosis and is suitable for use in the study of cardiovascular diseases.
- Reduces plasma cholesterol
- Reduces triglyceride levels
- Slows atherosclerosis progression
- Useful in cardiovascular disease studies