Learn More
Chemglass Life Sciences 70 μL Automation Clear Tips for Agilent System - Sterile, 384/rack (CLS-3499A-702)
SDP
Supplier:  Chemglass Life Sciences CLS3499A702
Encompass_Preferred
These 70 μL automation clear tips are sterile, low-retention pipette tips designed for high-throughput automated liquid handling workflows. Manufactured from medical-grade polypropylene and electron beam sterilized, they are DNase/RNase-free and intended for use with Agilent automated systems in genomics, proteomics, and other sensitive applications.
  • 70 μL nominal volume.
  • Low-retention surface to improve recovery of viscous liquids.
  • Sterile, electron beam sterilized and DNase/RNase free.
  • Packaged 384 tips per rack, 10 racks per pack, 50 racks per case.
  • Made from medical-grade polypropylene suitable for automated systems.
Catalog No. 50-273-3224