Learn More
Chemglass Life Sciences 70uL Automation Low-retention Clear Tips for Agilent System, Sterile, 384/rack, 10 racks/pk, 50 racks/cs
SDP
Supplier:  Chemglass Life Sciences CLS3499A602
Encompass_Preferred
NEST automation tips can be used for research and development and other commonly used applications with needs of high-throughput pipetting in genomics, proteomics, cytomics, immunoassays, metabolomics, biopharmaceuticals. Compatible with Agilent Systems.Low-retention tips are and excellent choice for transferring viscous liquid, which can effectively reduce the phenomenon of liquid hanging on the wall and make the experiment more accurate.• Medical-grade PP • Dnase/Rnase free • Electron beam sterilization • Produced in 100,008 grade clean room
Catalog No. 50-273-3228