Learn More
Refrigiwear Inc. 7120R NAVY/BLACK REG 3XLARGE FROSTLINE INSULATED BIB PANT
SDP
Supplier:  Refrigiwear Inc. 7120RNBK3XL
Encompass_Preferred
When you have to do precision work in subzero temps, bulky insulated overalls and coveralls can make that hard. Luckily, you can reach for these bibs and get to work without feeling weighed down. Frostline® Bib Overalls feature AirBlaze® insulation made from eco-friendly, sustainable materials that feel 40% warmer than traditional insulation. When workers aren't weighed down, they accomplish amazing things. With Frostline® Collection, you get incredibly lightweight gear that is durable, flexible and crazy warm. Comfort Rating: -25°F NEW! Improved fit and comfort 200g AirBlaze® insulation 100% Ripstop nylon outershell 100% Polyester lining Water-repellent, wind-tight finish YKK® zipper closure Hem & heel reinforcement patches Abrasion-resistant patches at knee and inseam Bound seams to seal out drafts Above-knee leg zippers 1 Zippered chest pocket 2 Snap-closure cargo pockets Adjustable straps for better fit Elastic waist back    
Catalog No. NC3886000